Hospira Plum A+ Infusion Pump
Supplier: GUMBO MEDICAL
HPAIP
470354-720EA 1307.69 USD
470354-720
Hospira Plum A+ Infusion Pump
Pumps Syringe Pumps
Scalable and programable.
  • Refurbished
The Hospira Plum A+ Infusion Pump is a scalable platform that can be initially deployed, and later upgraded to include Hospira MedNet safety software to meet evolving patient care needs. The unique, innovative PlumSet cassette technology and time-saving features simplify medication delivery and help to enhance safety for patients, clinicians, and caregivers. The Plum A+ is a proven, general-purpose pump that has been referred to as a work horse for its ability to provide effective concurrent IV infusion delivery.
This Hospira Infusion Pump technology allows users to deliver infusions with confidence and to limit medication errors. Many programming options are available with automated piggyback and concurrent delivery, programmable standby settings, multistep delivery, loading dose automation, and programmable delayed starts. The Plum A+ is designed to be used in most areas of patient care including general floor, medical/surgical, ICU, pediatrics, emergency, and much more.
Ordering information: Additional features and models may be available. Please contact your sales representative or customer service for information. This is a large item and will require freight shipment. Additional shipping may apply.
Caution: This item is refurbished and is intended for educational and training use only.