UniStore™ Sterile Media Bottles, PETG
Supplier: United Scientific Supplies
BMPETG1000 BMPETG125 BMPETG2000 BMPETG250 BMPETG30 BMPETG500 BMPETG60
470354-616CS 234.78 USD
470354-616 470354-618 470354-620 470354-622 470354-624 470354-626 470354-628
UniStore™ Sterile Media Bottles, PETG
Bottles Plastic Bottles
Durable and leakproof PETG sterile media botttles.
  • Manufactured in a Cleanroom Class 100K facility
  • Bottle materials meet USP Class VI requirements and are non-cytotoxic, non-hemolytic, and non-pyrogenic
  • Gamma sterilized with a Sterility Assurance Level (SAL) of 10⁻⁶
  • Bottles are leakproof tested and have a heat-shrink band around closure for tamper-evident seal
  • Temperature range: −40 to 70 °C
UniStore™ polyethylene terephthalate glycol (PETG) bottles are designed for long-term storage of media, biological buffers, and various reagents. Bottles are leakproof, durable, as well as freezer-safe, making them ideal for storage and transport. Includes a high-density polyethylene (HDPE) closure with heat-shrink band for a tamper evident seal. Bottles feature raised graduations for easy readability. Bottles are suitable for frozen storage to −40 °C. Bottles are Gamma sterilized and packaged in shrink wrapped trays within an outer bag.