Celestron Origin Intelligent Home Observatory
Supplier: Celestron International
12099
470354-444EA 3999.99 USD
470354-444
Celestron Origin Intelligent Home Observatory
Telescopes
Celestron Origin is an intelligent, all-in-one home observatory that captures stunning views of faint, deep-sky celestial objects and delivers them to a phone or tablet.
  • Revolutionary RASA optical technology
  • Automatic, AI-powered astrophotography
  • Automated setup with StarSense technology
  • Intuitive Celestron Origin app interface
  • Share the view with everyone; multiple users can use the Origin app simultaneously
  • Weight: 41.6 lbs.
Celestron Origin stands at the forefront of a new era in amateur astronomy, blending stargazing and astrophotography into a single, user-friendly experience. This intelligent, all-in-one home observatory takes the complexity out of traditional telescopes, transforming the backyard into a gateway to the cosmos. Packed with cutting-edge technology, Celestron Origin captures the beauty of celestial objects and brings them to life on a phone or tablet.
Origin’s patented 6" Rowe-Ackermann Schmidt Astrograph (RASA) optical design places it in a class by itself. With its large aperture and fast f/2.2 focal ratio, you get unparalleled image brightness and clarity with less exposure time. As Origin captures data, built-in AI algorithms autonomously stack and process every frame in real-time. Galaxies and nebulae instantly come to life - sharp, detailed, and in brilliant color - all with no user intervention.
To get started, place Origin outside, turn it on, and connect to the app. Using Celestron’s patented StarSense technology, Origin scans the sky and autonomously focuses and aligns itself. You’re ready to select a target and start imaging in less than two minutes. The user-friendly Celestron Origin app for iOS and Android, developed in partnership with Simulation Curriculum, the makers of SkySafari™, offers a simple yet powerful planetarium interface, making it easy to navigate the sky and control the system. Multiple users can use the Origin app to view the telescope’s live feed simultaneously. You can also cast the view to a smart TV or save and share it instantly. As you grow in the hobby, download the raw image files you capture for more advanced manual processing. Also experiment with filters and use Origin for night-vision or land-based observing during the day.
Celestron engineers drew on their decades of expertise while designing every detail, from Origin’s patented optical technology to its intuitive companion app. The result is a revolutionary new system that builds on Celestron’s legacy and makes astronomy more accessible, exciting, and fun than ever before.