Knife Switch, Small
Supplier: United Scientific Supplies
KSW001
470353-956EA 2.55 USD
470353-956
Knife Switch, Small
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Tradiitonal lever switch for circuit experiments.
  • Easy pivot lever
  • Secure connection screws
Single-pole, single-throw knife switch with plastic base is suitable for low voltage experiments (less than 3.6 V). Includes thumbscrew connecting posts.