Streak Plates, Glass
Supplier: United Scientific Supplies
GLP1X2-P
470353-570EA 1.2 USD
470353-570
Streak Plates, Glass
Staining Tiles
Clear glass for mineral testing.
  • ¹/₄" thick glass
  • Overall dimensions: 2×1"
Designed for use in hardness testing of rocks and minerals.
Packaging: Plates are sold as packs of 10.