Disposable Borosilicate Glass 7.0 Pasteur Pipets
Supplier: United Scientific Supplies
DBG146NS DBG146NSCP DBG229NS DBG229NSCP
470353-238PK 118.54 USD
470353-238 470353-234 470353-286 470353-284
Disposable Borosilicate Glass 7.0 Pasteur Pipets
Pipettes Pasteur/Transfer Pipettes
Disposable Pasteur pipettes are ideally suited for use in both clinical and industrial laboratories.
  • Manufactured from Borosilicate glass 7.0
  • One-time use eliminates the danger of cross-contamination
  • 2 ml capacity
  • Available in plain or cotton plugged options
Disposable Pasteur pipettes provide an efficient means for inoculating culture media and transferring samples and reagents in both clinical and industrial laboratories. These 2 ml capacity, non sterile, and disposable pipettes are manufactured from borosilicate glass 7.0. Available in two sizes, with or without cotton plugs.