Ward's® Glider Launcher
Supplier: United Scientific Supplies
GLDLCHRVWR
470353-040EA 98.38 USD
470353-040
Ward's® Glider Launcher
Educational Materials Physics Educational Materials Mechanics Learning Activities
Versatile design to launch a variety of rubber band powered gliders.
  • Sturdy base
  • Built-in angle indicator
  • Long 24" ramp
This glider launcher is loaded with different features to enhance classroom activities. The long 24" base provides plenty of room to pull back the glider before release. A screw on the end of the ramp securely holds a rubber band in place. A wing nut and bolt allows the entire ramp to be angled and then positioned at a desired angle.
The launcher is equipped with a protractor on the side that reads out degrees from 0 to 90 degrees. A metal arm pivots freely as the ramp is adjusted so the exact angle can be determined. Required but not included are gliders and rubber bands.