Skin Suture Trainer
Supplier: American 3B Scientific
1024494 1024495
470352-886EA 199.2 USD
470352-886 470352-892
Skin Suture Trainer
Models Human Biology Models
Practice suturing on either a flat or curved surface.
  • Practice multiple suture methods
  • Includes a base to provide tension on the suture pad
  • Two skin tones available
The skin suture trainer by 3B Scientific perfectly replicates the anatomical structure of human skin layers including the epidermis, dermis, and hypodermis, to create a realistic haptic experience of touch for the trainee. By using the included housing base, users can provide skin tension to the suture pad for an even more realistic suture experience. This skin tension simulates the direction of orientation of the underlying collagen fibers. With a sharp scalpel, wounds can be cut to match the required depth and location on the pad. Suture and staple material can be removed from the incisions allowing for the trainer to be reused for many training exercises. The material is very robust for dependably high-quality, long-term use.