Eisco Microcentrifuge Tube Float Rack
Supplier: Wards
CH0285PK12
470352-730CS 829.6 USD
470352-730
Eisco Microcentrifuge Tube Float Rack
Racks Tube Racks
Economical Retort Bases with tapped rod hole made of powder-coated pressed steel.
  • Racks hold 16 microcentrifuge tubes with a 1.5 ml capacity for incubation in water baths
  • Polypropylene construction for strength, buoyancy, and temperature resistance
  • Autoclavable
  • Floats when fully loaded with filled tubes
  • Surface of the rack has raised alpha-numeric index for easy identification
  • Central rod handle for convenient lifting
  • Measures 95×95 mm
Float rack for suspending 1.5 ml microcentrifuge tubes in water baths. The easy-to-load 16-place rack is made of polypropylene which is buoyant, durable, and autoclavable. An essential equipment for microbiology, biotechnology, and immunology labs.