Cubelets® Boundless Builders Pack
Supplier: MODULAR ROBOTICS
855165004840
470352-686PK 699.99 USD
470352-686
Cubelets® Boundless Builders Pack
Educational Materials Physics Educational Materials Applied Physics Learning Activities
A full set of Cubelets® for a home or makerspace.
  • Designed to be intuitive and screen-free for the youngest learners
  • Incorporate coding and computer science principles for older students
  • Kit supports one student group
  • Includes 19 Cubelets® (16 of 16 Cubelet types), and 4 brick adapters
  • Neatly stores away in one durable plastic tub
A little bit of everything. The Boundless Builder gives you access to a full set of Cubelets® for the best screen-free robot building experience. It's the perfect way to introduce STEM, robotics, and computational thinking. Foster vital thinking skills in your classroom, makerspace, library, or museum. Fascinating and fun, Cubelets® nurture your student's imagination, creativity, and inventive spirit. Cubelets® are the building blocks of better computational thinkers.
Assembled for eager inventors, the Boundless Builder includes 19 SENSE, THINK, and ACT Cubelets®. Designed to support a single student group, this pack has tons of building options as students complete challenge after challenge. Also included are brick adapters, a micro-USB charging cable, and durable storage. Every Cubelets® set and pack includes access to free standards-aligned lesson plans, printable activity cards, online professional development, and more.
Cubelets® magnetic faces make them simple to snap together and pull apart. Each Cubelet has a special function. What you connect, and how you connect them changes the behavior of your robot constructions: every combination is a new creation. Each Cubelet belongs to one of three categories: SENSE Cubelets® take in information from the robot's environment. THINK Cubelets® compute and change that information, and ACT Cubelets® turn that information into physical action. Connect at least one SENSE, THINK, and ACT Cubelet to create a robot construction. Inside of each block is a part of a program – as you build your robot you build the code that controls its behavior. It's hands-on coding.
Ordering information: Recommended accessories: (1) bluetooth hat.
Delivery information: Kit includes 19 Cubelets®:
(1) brightness, (2) distance, (1) battery, (1) inverse, (1) speaker, (1) temperature, (2) drive, (1) rotate, (1) flashlight, (1) maximum, (1) passive, (1) blocker, (1) threshold, (1) bar graph, and (1) minimum. Plus: (2) stud brick adapters, (2) socket brick adapters, (1) charging cable and (1) plastic storage tub.