Copper Strips
Supplier: Avantor
470352-650PK 17.99 USD
470352-650
Copper Strips
Specimens Mineral Specimens Classroom Minerals
Great for geology hardness tests.
  • Pure copper
  • Cut to size for mineral and rock ID testing
  • 0.1 mm thickness