Trundle Wheel
Supplier: United Scientific Supplies
TRUNWHL
470352-574EA 65.03 USD
470352-574
Trundle Wheel
Measuring Tapes
Easily measure out long distance.
  • Measures up to 99,999 meters
  • Displays in increments of 0.2 m
  • Wheel has 1 cm markings along the edge
This trundle wheel easily rolls along the ground to measure linear distance. The rubber wheel edge provides a steady grip when rolling over different surfaces. Telescoping handle can extend from 43 to 76 cm to provide comfortable operation for all users. The built-in folding stand allows upright storage.
Delivery information: Includes a convenient carrying case.