HOBO® TidbiT MX External Temp Data Logger
Supplier: ONSET COMPUTER CORPORATION SE
MX2205
470352-230EA 189.44 USD
470352-230
HOBO® TidbiT MX External Temp Data Logger
Data Loggers
Bluetooth logger with external sensor.
  • Fast temperature-response time (20 sec.)
  • External temperature sensor, plus internal sensor for ambient temp monitoring
  • ±0.2 °C (±0.36 °F) accuracy
  • Large memory stores 96,000 measurements
  • 3-year battery life (Bluetooth on); 5-year battery life (Bluetooth off)
  • LED indicates when temperature exceeds set threshold
The HOBO TidbiT MX2205 is ideal for monitoring shallow waters, like streams and wetlands, or for monitoring soil in fields that are prone to flooding. Equipped with both an internal sensor to monitor ambient temperature and an external sensor on a 2 m cable to accurately monitor fast-changing temperatures, the HOBO TidbiT Mx2205 is a low-cost, easy-to-use, waterproof logger that leverages the power of Bluetooth to deliver high-accuracy data straight to your mobile device or Windows laptop running Onset's free HOBOconnect app. This rugged logger withstands harsh environments, and can be deployed just above water or soil for quicker, more convenient data downloads that don’t require pulling up the logger.
Ordering information: Requires a compatible mobile device or Windows computer and the HOBOconnect app.