Plastic Dropper
Supplier: United Scientific Supplies
PLPIP3
470352-216PK 44.7 USD
470352-216
Plastic Dropper
Pipettes Pasteur/Transfer Pipettes
Economical plastic dropper.
  • PP tube
  • Rubber bulb
Straight plastic pipettes with a capcity of 1 ml. The tips are PP and the bulb is made out of rubber.