miniPCR® Biotech Classroom Pack
Supplier: AMPLYUS LLC
QP-2500-28
470352-076EA 7707.09 USD
470352-076
miniPCR® Biotech Classroom Pack
Educational Classroom Kits and Activities
Bring cutting-edge biotechnology into your classroom with this Biotech Classroom Pack.
A versatile option for classes between 16 and 64 students working in 4 to 16 lab groups.
Certifications: The miniPCR® thermal cyclers and blueGel electrophoresis systems are CE marked, made in the USA, and backed by the miniPCR 2-year Warranty and 30-day satisfaction guarantee!
Delivery information: This Biotech Classroom Pack includes: 4 miniPCR® mini8X thermal cyclers: Simple and intuitive, the mini8 offers a truly personalized PCR experience for maximum engagement. 8 blueGel™ electrophoresis systems: Fast, durable, safe, and easy to use, blueGel™ allows you to visualize results instantly in a convenient, all-in-one device compatible with safe DNA dyes such as GelGreen®. 2 Gyro™ Microcentrifuges: This robust, single speed microcentrifuge is user friendly and includes rotors for both 1.5 ml tubes and 0.2 ml tubes. Micropipetting supplies: Our H-style micropipettes are durable, accurate, and fully adjustable. Pack includes: 8 student micropipettes (2 to 20 μl), 1 teacher set of 3 micropipettes (0.5 to 10 μl, 2 to 20 μl, 20 to 200 μl), and compatible tips (8 racks 200 μl tips, 2 racks 10 μl tips).