miniPCR® Learning Lab Companion Kit
Supplier: AMPLYUS LLC
KT-1510-01
470352-026EA 45.09 USD
470352-026
miniPCR® Learning Lab Companion Kit
Educational Classroom Kits and Activities
A handy bundle of all the consumables you need to complete a miniPCR bio Learning Lab with a class of 32 students.
SeeGreen™ All-in-one Agarose Tabs™ features:
- All-in-one tabs: SeeGreen™ stain and TBE buffer included
- Convenient format - no weighing required
- Fast dissolving - gels ready to pour in seconds
- High transparency and low background
- Stable at room temperature for 3 years
Please note:
This item can also be ordered as an add-on to miniPCR Learning Labs at no additional shipping cost. See Learning Lab product pages for more information. Micropipette tips not included
Delivery information: Includes 50 microtubes (1.7 ml); 100 PCR tubes (0.2 ml); 5.1 g vial of TBE electrophoresis buffer powder (sufficient to make 600 ml of 1X TBE buffer); 8 SeeGreen™ All-in-one Agarose Tabs™ sufficient for 8 gels (2%) or up to 16 gels (1%) for the blueGel™ system.