Estes Porta Pad E Launch Systems
Supplier: Estes Industries
2230
470351-954EA 42.4 USD
470351-954 470351-956
Estes Porta Pad E Launch Systems
Educational Materials Physics Educational Materials Mechanics Learning Activities
The Porta pad E launch system is large enough to accomodate those rockets that require larger, 24 mm E and F engines. It has a larger footprint for added stability and a larger blast deflector plate. The Porta pad E comes with the larger ³/₁₆" Maxi launch rod.
  • Porta pad E: ³/₁₆" two-piece Maxi launch rod assembles to 80 cm (31.5")
  • E controller features 9.1 m (30') launch cable and requires (4) AA batteries
With a long, 30' cable, the E launch controller is perfect for even the largest Estes model rockets. The four AA-batteries provide plenty of electric current needed to ignite your Estes model rocket engines, and the long cable keeps young rocketeers at an even safer distance. A safety key lockout system ensures that electricity cannot flow while attaching and adjusting the leads to the Estes STARTECH engine starter (igniter).
Rocketeers should always wear Personal Protective Equipment (PPE), and carefully ensure that all personell are outside the 30' launch radius before inserting the key and pushing the launch button.
Ordering information: The launch pad and controller must be ordered separately. Required but not included: (4) AA batteries.
Delivery information: Porta pad E includes (1) tripod launch pad, (1) blast deflector, (1) two-piece launch rod, instructions. Launch controller E includes (1) launch controller with 30' cable, (1) safety key, instructions, safety code, one-year warranty.