ACCU-SCOPE® C-Mount Microscope Camera Adapters
Supplier: ACCU-SCOPE INC SE
00-2010-1 00-2010-35 00-2010-50 00-2010-65 00-2010-P
470351-132EA 144.85 USD
470351-132 470351-246 470351-120 470351-254 470351-122
ACCU-SCOPE® C-Mount Microscope Camera Adapters
Adapters Adapters, Camera to Microscope
Microscope camera adapters are designed to fit ACCU-SCOPE's EXC-400, EXC-350, 3001-LED or 3001 microscopes.
  • Multiple sizes available
Adapter can be focused to match the focus of the digital image to the image seen through the eyepieces.