ACCU-SCOPE® 3000-LED Upright Microscope Series
Supplier: ACCU-SCOPE INC SE
3000-LED 3001-LED 3001-LED-6HDS 3001-LED-HD 3001-LED-SPH
470351-110EA 1757.8 USD
470351-110 470351-166 470351-262 470351-276 470351-082
ACCU-SCOPE® 3000-LED Upright Microscope Series
Microscopes
Delivers excellent optical performance, value, and resolution to meet the exacting standards for life science professionals and students.
  • Binocular or Trinocular viewing heads
  • Head inclined 30°, rotatable 360°
  • HWF Plan 10×/20 mm focusing eyepieces (paired - one with pointer), with eye guards
  • Reversed quadruple nosepiece Infinity Plan achromat 4×, 10×, 40×R and 100×R oil objectives
  • Mechanical stage
With a best-in-class 20 mm field of view and super-bright LED illumination, the 3000-LED Series produces high contrast images with outstanding resolution. Our trinocular 3001-LED models accept digital cameras for documentation, instruction, and even greater ergonomics.