Digital Rotator
Supplier: LW Scientific
RTL-BLVD-24T3
470351-010EA 658 USD
470351-010
Digital Rotator
Shakers and Mixers Rotating/Rocking Shakers
The Serology rotator is designed to give years of dependable service, even in the busiest laboratory. It is the ideal space-saving instrument for procedures that require constant or timed rotation (mixing) such as RPR, VDRL, latex tests and for procedures that require a nonstandard speed.
  • Widely used in VDRL, RPR, and EIA tests
  • Stable and quiet operation
  • Clear humidity tray
  • Variable speed, timer control and continuous operation
Load capacity: 4.4 lbs. at 20 to 150 rpm, 2.2 lbs. at 151 to 240 rpm
Orbit diameter: 0.8"
Delivery information: *Includes humidity cover.