Microhematocrit Centrifuge
Supplier: LW Scientific
MHC-24HD-75T1
470351-008EA 1525.3 USD
470351-008
Microhematocrit Centrifuge
Centrifuges Microcentrifuges
This digital, 24-place microhematocrit centrifuge comes with a reader disk.
  • Digital controls
  • Adjustable speed and time
  • EZ Reader microhematocrit card included
Designed and assembled in the USA, this centrifuge features a powerful, maintenance-free brushless motor that spins 75 mm microhematocrit tubes in under three minutes. Then read the packed red cell volume using the included reader disk. Sturdy all-metal construction, and suction cup feet keep this compact unit stable on the countertop.