Extech® Waterproof Palm pH Meter with Temperature
Supplier: NEW GENIE GROUP INC SE
PH220-C
470350-914EA 198.5 USD
470350-914
Extech® Waterproof Palm pH Meter with Temperature
Electrochemistry Meters pH and Multi-Parameter Meters
This palm-sized pH meter with cabled electrode can measure pH, voltage potential difference (mV) and temperature.
  • Internal memory stores up to 25 readings
  • Built-in microprocessor provides for automatic buffer recognition
This completely waterproof unit features a large LCD display with simultaneous measurement readings and low battery indicator.
Delivery information: Includes cabled pH electrode, protective holster, pH buffers, and 9 V battery.