Isoamyl Benzoate Thermometers
Supplier: United Scientific Supplies
UNTRMABDS UNTRMPM12
470350-692EA 2.55 USD
470350-692 470350-690
Isoamyl Benzoate Thermometers
Thermometers Analog Thermometers
A safer option for measuring temperature.
  • Non-toxic filling (isoamyl benzoate) is safer than mercury and other spirits
  • Bright blue fluid against a white background makes for easy reading
  • Should be stored in an upright position to prevent the column from separating
Partial immersion thermometers are useful if you have a shallow container which holds the substance to be measured. These thermometers have a small indicator ring that denotes the depth to which the thermometer should be submerged to obtain an accurate temperature reading. The blue isoamyl benzoate is a non toxic liquid and easier to read. The thermometers features a dual scale in Fahrenheit and Celsius.