Hanna Backpack Lab Water Quality Educational Test Kit
Supplier: HANNA INSTRUMENTS
HI3817BP
470349-864EA 541.83 USD
470349-864
Hanna Backpack Lab Water Quality Educational Test Kit
Educational Materials Environmental Educational Materials Environmental Learning Activities
Classrooms can gain an extensive understanding of water quality through measurement of ten parameters.
  • Students will asses the water quality of streams, rivers, and lakes
  • Includes portable test kits, pocket testers, lessons, and actvities
A classroom in a backpack! The Backpack Lab Water Quality Educational Test Kit contains lessons, activities, and test kits to help students understand the importance of common water quality parameters.
Using titration, colorimetry, and direct measurement, educators and students can easily measure parameters such as acidity, alkalinity, carbon dioxide, dissolved oxygen, hardness, pH, EC, TDS, and turbidity. Supplied with all of the necessary reagents and equipment to perform each analysis, and all reagents and buffers are individually available as they run out.