Dedicated pH Meter
Supplier: HANNA INSTRUMENTS
HI2002-01
470349-852EA 479.31 USD
470349-852
Dedicated pH Meter
Electrochemistry Meters pH and Multi-Parameter Meters
This pH meter is seriously thin and light with lab grade functionality packed into a tablet design. With simple operation and advanced features, edge pH easily outperforms all other portables and benchtops in its class. Advanced pH electrode diagnostics ensure worry-free measurements. Use this pH meter as a portable, benchtop meter, or mount it on the wall to save space.
  • Stunning 5¹/₂" screen with 150° viewing angle provide one of the easiest to read LCDs in the industry
  • Built for everyone from educators and researchers to home users
Wall Mount-Using the wall mount cradle (included), edge can be placed on a wall, leaving zero footprint on the benchtop space. The cradle has a built-in connector to power the meter and charge its batteries. edge's zero footprint is designed to save you valuable benchtop space.
Cradle and Electrode Holder- This meter is equipped with a benchtop cradle with an adjustable swivel electrode holder to charge and hold the meter securely in place at the optimum viewing angle.
Data Logging- this digital pH meter allows you to store up to 1000 log records of data. Data sets include readings, GLP data, date and time.
Ordering information: In order to enable ORP functions you would need to purchase the ORP Probe sold seperately. pH calibration solutions, storage solutions, and cleaning solutions will need to be purchased separately. Contact your sales representative for more information.
Delivery information: Includes digital pH meter, a smart pH electrode, buffer and cleaning solution sachets (2 each), bench cradle with electrode holder, wall mount, adapter.