Hanna Instruments NPK Soil Chemical Test Kit
Supplier: HANNA INSTRUMENTS
HI3895
470349-848EA 41.67 USD
470349-848
Hanna Instruments NPK Soil Chemical Test Kit
Environmental Test Kits
This chemical test kit will determine the commonly monitored parameters in soil testing.
  • Pre-made reagents for ease of use
  • Measure pH for the most important elements of plant growth: Nitrogen, phosphorus and potassium
  • All reagents marked with expiration date and lot number for traceability
  • Test tubes for convenient handling
This test kit is supplied with all of the necessary reagents and equipment to perform each analysis, and all reagents are individually available as they run out. It provides users with a fundamental tool to assess important characteristics related to soil quality. All required materials are included with the test kit, such as the test tubes, color comparison cards, reagents and solutions. Methods include colorimetric for pH and nitrogen and turbidimetric for potassium.
Ordering information: This kit is equipped with all the necessary reagents and equipment to perform over 10 tests for each parameter. Replacement reagents sold separately.