Multiparameter pH meter
Supplier: HANNA INSTRUMENTS
HI2020-01
470349-840EA 546.81 USD
470349-840
Multiparameter pH meter
Electrochemistry Meters pH and Multi-Parameter Meters
This laboratory pH meter delivers precision while offering simplicity and versatility in a sleek design.
  • The large display and it's wide 150° viewing angle provide one of the easiest to read LCDs in the industry
  • Setup and calibrate your meter in no time with as little as a 1 point calibration
  • Great for use in business and educational laboratories worldwide
Your electronic pH meter will automatically recognize each digital electrode, providing sensor type, calibration data, and serial number. Advanced pH electrode diagnostics ensure worry-free measurements. Use your multiparameter pH meter as a portable, benchtop meter, or mount it on the wall to save space.
Wall Mount-Using the wall mount cradle (included), edge can be placed on a wall, leaving zero footprint on the benchtop space. The cradle has a built-in connector to power the meter and charge its batteries. edge's zero footprint is designed to save you valuable benchtop space.
Cradle and Electrode Holder- This meter is equipped with a benchtop cradle with an adjustable swivel electrode holder to charge and hold the meter securely in place at the optimum viewing angle.
Data Logging- this digital pH meter allows you to store up to 1000 log records of data. Data sets include readings, GLP data, date and time.
Ordering information: Dissolved oxygen and conductivity electrodes purchased seperately to enable you to measure conductivity, total dissolved solids (TDS), and dissolved oxygen. Calibration solutions purchased seperately. pH storage and cleaning solution purchased seperately. pH electrode sold seperately. Contact your sales representative for more information.