Equipotential Trough Kit
Supplier: United Scientific Supplies
UNEQPTK01
470349-678EA 52.99 USD
470349-678
Equipotential Trough Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Plot equipotential lines accurately with water and a digital multimeter.
  • Plot equipotential lines for various electrode configurations
  • Classic design
  • Correlates to NGSS Standards MS-PS2-5 and HS-PS2-4
  • Activity guide included
This comprehensive kit allows students to plot equipotential lines in an electric field between two electrodes held at different electric potentials. A simple setup keeps electrodes partially immersed in water containing an electrolyte, allowing a small current to flow between the electrodes. Students can then sample the resulting electric field using the included multimeter.