Alnico Bar Magnets
Supplier: United Scientific Supplies
ABM020
470349-636PK 9.99 USD
470349-636
Alnico Bar Magnets
Magnets
Strong Alnico bar magnets for demonstrations.
  • Includes a keeper
  • Strong Alnico magnets
  • Perfect for demonstrating magnetic fields
Powerful Alnico 1 bar magnets, shipped in pairs, with end keepers. Magnets are painted with North and South poles, clearly marked. Each bar is 2" long and ¹/₄" thick.