Solar Motion Model
Supplier: Science First
10240
470349-334EA 255 USD
470349-334
Solar Motion Model
Models Astronomy Models
Clearly shows the path of the sun at any latitude during any month of the year.
  • Elegant, easy to use, and hands-on model
  • Wooden desk top model with clean finish
  • Stands 13½" high
Ever wonder, 'What is the path of the sun where you live on Dec 31 or June 15?' This device will teach students how this astronomical feature is calculated.