Crazy Circuits™ Coding Kit
Supplier: BROWN DOG GADGETS
K1049
470348-930EA 114.4 USD
470348-930
Crazy Circuits™ Coding Kit
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Combine the fun and creativity of brick-based building with the power of Arduino programming.
  • LEGO™ brick-compatible programming platform powered by Arduino
  • Designed to teach the fundamentals of robotics and coding
  • No soldering or additional tools needed
Welcome to the world of programming and automation with Arduino™. The Arduino-powered robotics board allows for the easy addition and programming of included sensors and parts without the need for a breadboard or soldering. Crazy Circuits™ parts are compatible with LEGO™ and similar brick building systems. Crazy Circuits™ parts come 100% ready to go out of the box, which means no soldering or preparation required. This kit covers topics such as circuitry, programing, logic, art, and design. These modular kits are robust sets of materials supported by online lesson plans, projects, activities, diagrams, and videos. This is essentially an Arduino™ version of our bit board kit for those with a preference for that platform over micro: bit™.
Delivery information: Kit includes (4) 10 mm LEDs, (4) 1×6 LEGO™ pieces, (2) small pushbuttons, (1) slide switch, (1) Neopixel, (1) potentiometer, (1) piezo speaker, (1) Crazy Circuits™ Robotics Arduino™ board, (1) roll ¹/₈" × 5 m Maker Tape™, (1) USB cable, (1) bundle of 5 alligator clips, (1) LEGO servo + connector piece, (1) LEGO™ base plate, instruction cards/online curricula.