miniPCR® P51™ Molecular Fluorescence Viewer
Supplier: AMPLYUS LLC
QP-1900-01
470348-600EA 40.99 USD
470348-600
miniPCR® P51™ Molecular Fluorescence Viewer
Transilluminators Blue Light Transilluminators
Visualize DNA, RNA, and proteins at work right in the palm of your hand.
  • Tube holder design allows for pipetting while tubes are in the viewer
  • Views 8 tubes at once
  • Power source either through 9V battery or USB-C
  • Includes orange filter for performing BioBits and P51 Molecular Glow Labs
  • Bonus yellow filter for enhanced viewing of yellow and green fluorescence
With P51™, you can detect and measure molecules like DNA, RNA and proteins in a simple, visual way. Scientists use fluorescence every day to detect and quantify DNA, RNA, and proteins, but this often requires sophisticated and complex equipment. With P51™, we have packaged the power to visualize molecules into a compact, cheap, handheld format that’s perfect for any setting.
Through hands-on investigations, students will explore the building blocks of life by visualizing them in the P51™ Fluorescence Viewer. Simply place your samples inside the P51™ and turn on the safe blue light – and see your tubes light up!