miniPCR® GELATO™ Electrophoresis and Visualization System
Supplier: AMPLYUS LLC
QP-1600-01
470348-556EA 1023.09 USD
470348-556
miniPCR® GELATO™ Electrophoresis and Visualization System
Electrophoresis Systems Vertical Gel Electrophoresis Systems
Electrophoresis for scientists with great taste™.
  • Ultra-fast, high-capacity system - run up to 50 lanes at a time
  • Built-in power supply with adjustable voltage settings 50 to 135 V
  • Integrated transilluminator for real-time DNA visualization
  • High-intensity LED panel for eye-popping bands
  • Safe blue light compatible with safe nucleic acid stains including SeeGreen™, GelGreen®, SYBR®
  • Direct on-bench image capture
  • Fold-a-View™ photo documentation hood included - a portable, foldable darkroom!
  • Orange goggles and cutting tray included
GELATO™ integrates gel electrophoresis and a blue light transilluminator so you can run, view, snap, and cut bands - all in one system! With the GELATO™ system, you can separate nucleic acids at ultra-fast speed, instantaneously view eye-popping bands, snap pictures and cut bands right on your bench. All in one device! Get publication-quality results in a compact, space-saving footprint.