REUZ™ Stainless Steel Beakers
Supplier: United Scientific Supplies
UN3007-100 UN3007-1000 UN3007-2000 UN3007-250 UN3007-3000 UN3007-500 UN3007-5000
470347-910EA 23.1 USD
470347-910 470347-924 470347-972 470347-986 470347-914 470347-980 470347-930
REUZ™ Stainless Steel Beakers
Beakers
Seamless and sanitary stainless steel beakers.
  • 304 stainless steel
  • Reusable and long-lasting
  • Easy to clean
  • Seamless, single piece construction
  • Available in sizes from 100 to 5000 ml
Impact-resistant and built to last, United Scientific REUZ™ ('re-use') beakers are made of 304 stainless steel. The precision corrosion-resistant stainless construction includes rolled top rims, smooth edges and radiused corners for easy, thorough cleaning. These beakers do not contain graduation markings - designed to be filled to the brim to realize full vessel capacity. Useful for storing, mixing, and heating solutions in laboratory and production settings when durability and easy sterilization is desired. Wall thickness is 1 mm.
REUZ™ products promote sustainability in all laboratory environments.
Ordering information: Sold individually.