Zener Diode
Supplier: United Scientific Supplies
DIZEN500MWPK10
470347-906EA 2.8 USD
470347-906
Zener Diode
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Control the current direction in circuits.
  • 2 pin connectors
  • Fits all standard bred boards
Zener diodes are a unique electrical component that allows the current to flow in a “reverse direction”. A diode only allows a current to flow in one direction and prevents an electrical charge being reversed. A Zener diode does allow the current flow to be reversed, but only once the voltage equals the specific voltage rating.