LoadWell™ Gel-Loading Pipette Tips
Supplier: MTC BIO, INC.
P5720
470347-460PK 128.2 USD
470347-460
LoadWell™ Gel-Loading Pipette Tips
Pipette Tips
These thin-tip pipette tips are ideal for precise sample dispensing into electrophoresis wells.
  • Narrow diameter, flexible capillary
  • Universal fir for standard pipettors
  • E-beam sterilized
  • Free of RNase, DNase, and pyrogens
  • Convenient rack packaging
LoadWell™ Gel loading tips have a 0.7 mm outer diameter microcapillary orifice, ideal for introducing samples into gel electrophoresis wells. Designed to fit all standard 200 µl pipettes (as well as 20, 50 and 100 µl pipettes), they are supplied sterile (e-beam), and packaged in autoclavable racks of 96 tips. Produced from virgin, medical grade polypropylene, they are RNase, DNase, endotoxin, and pyrogen free.