Innovating Science® Total Water Investigation
Supplier: ALDON CORP SE
IS9763
470347-440EA 253 USD
470347-440
Innovating Science® Total Water Investigation
Environmental Test Kits
Easy way to test water for multiple factors.
  • 10 tests for water samples
Test water sample for alkalinity, ammonia, biochemical oxygen demand, chloride, coliform bacteria, copper, hardness, iron, nitrate, dissolved oxygen, pH, and phosphate.
Delivery information: Includes 3 water collection bottles; 6 reaction vessels; 6 measuring tubes; 15 pipettes; 6×1 ml titrators; 2×3 ml titrators; 1 pack toothpicks; 10 phosphate powder pillows; 1 pack pH strips; 10 iron test strips; 10 Copper test strips; 5 g coliform test powder; 3×100 ml silver nitrate titrant; 3×15 ml salinity indicator solution; 3×60 ml EDTA titrant; 50 ml pH 10 buffer solution; 15 ml Erichrome black T solution; 60 ml Griess reagent; 0.5 g cadmium metal powder; 15 ml total alkilinity indicator; 15 ml phenolphthalein indicator; 3×60 ml hydrochloric acid titrant; 15 ml ammonia test solution 1; 15 ml ammonia test solution 2; 15 ml ammonia test solution 3; 2×30 ml starch indicator solution; 2×30 ml mangenese chloride reagent; 2×30 ml alkaline iodide reagent; 2×30 ml sulfuric acid, 50% and 2×60 ml sodoium thiosulfate titrant.