Swift M3603 Series Microscope and Camera Bundles
Supplier: Swift Instruments
M3603C-WF3-X5
470317-660EA 1023.3 USD
470317-660 470347-388
Swift M3603 Series Microscope and Camera Bundles
Microscopes
Now with built-in camera port, these bundles includes a digital camera and video C-mount adapter.
  • LED light rated up to 50,000 hours of life
  • Single 8 hour charge allows up to 50 hours of use
  • Built-in handle
With coaxial focusing and upgraded LED illumination, the M3603C models are the ideal student microscopes. Paired with a 2.0 megapixels Moticam 2 or a WiFi enabled Moticam X3 you can go from conventional to digital microscopy in seconds.