Welch Allyn® CP150 Compact Resting ECG
Supplier: Healthcare Products
WEL CP150A-1ENB WEL CP150AS-1ENB WEL CP150AW-1ENB WEL CP150AWD-1ENB WEL 410370 WEL 714730 WEL 719653 WEL 720705 WEL 105660 WEL 105343 WEL 105353 WEL 105410
470347-110EA 6752.72 USD
470347-110 470347-064 470347-176 470347-168 470347-114 470347-268 470347-248 470347-238 470347-280 470347-194 470347-118 470347-210
Welch Allyn® CP150 Compact Resting ECG
Data Loggers
Simple and easy to use design.
  • Durable and high-quality materials
  • 7" TPT Color-touch screen interface
  • Rechargeable battery can last up to 8 hours of continuous operation
The Welch Allyn® CP 150 Compact Resting Electrocardiograph is a simple and efficient 12-Lead EKG machine aimed to help medical professionals in diagnosing and monitoring heart conditions by recording the electrical activity of the heart at rest. This machine can provide information about heart rate and rhythm, and can detect problems such as heart enlargement, history of heart attack (myocardial infarction) and other conditions, which can greatly help in properly treating the patient.