Welch Allyn® Veterinary Diagnostic Set
Supplier: Healthcare Products
WEL 96220
470347-240EA 1483.56 USD
470347-240
Welch Allyn® Veterinary Diagnostic Set
Opthalmoscopes
Easily portable.
  • Ophthalmoscope, Otoscope, rechargeable handle and hard carry case
  • 60 minutes on battery operation
  • Coaxial Ophthalmoscope with 28 lenses and 18 aperture combinations
  • Pneumatic Otoscope with white light and rotating lens
  • Hard case for storage and transportation convenience
Welch Allyn Coaxial Ophthalmoscope features high standard optics and smart design. It comprises 28 lenses that ensure excellent vision with a resolution ranging from -25 to +40 diopters. Moreover, the device performance is enhanced with a coaxial optical system that provides a shadow-free spot vision and easier entry into undilated pupil. Crossed linear polarizing filter and red-free filter are applicable with any aperture. The Ophthalmoscope head has 6 apertures (micro, small, and large spot sizes, fixation target, slit aperture, and red-free filter) for all general and specialist uses, and 18 possible aperture combinations for greater versatility. The smart outer construction with sealed optics and rubber brow rest protect the device from dust and eyeglasses scratches.
The Pneumatic Otoscope provides bright, white light output via fiber-optic illumination and a rotating lens with ultra-seal for easy pneumatic otoscopy. The otoscope has two viewing lenses for dual observation of the tympanic membrane. The unit comes with three polypropylene specula.