Welch Allyn® Tycos DS 48 Aneroid Sphygmomanometer
Supplier: Healthcare Products
WEL 5090-02CB
470347-220EA 359 USD
470347-220
Welch Allyn® Tycos DS 48 Aneroid Sphygmomanometer
Blood Pressure Monitors
Unsurpassed reliability.
  • Mercury-free and latex-free for safety
  • Jeweled movement for long life
  • Laser-engraved dial face for accuracy
  • One-piece cuff is more durable and easier to use
Tycos DS48 Pocket aneroid sphygmomanometer with DuraShock™ gear free, super shock resistant technology, jewel movement, laser engraved dial, lifetime calibration warranty, premium inflation bulb and valve, zipper case, size 11 adult, reusable (two piece), 2-tube cuff (US only).
Provides protection to the manometer during the rigors of clinical use. Precision, jeweled bearings resist wear for consistently accurate readings. Gauge attached to cuff protects instrument and prevents cuff and gauge from being easily separated. Laser-engraved dial for precise calibration. Unique phosphor-bronze bellows ensure consistent pressure measurement. Crisp, white graphics on large charcoal-gray scale for better readability. Lifetime warranty on calibration and manufacturing quality. All latex free.