Welch Allyn® ELI 280 Resting ECG
Supplier: Healthcare Products
WEL ELI280-AAA-AAFBD
470347-170EA 16366.82 USD
470347-170 470347-196
Welch Allyn® ELI 280 Resting ECG
Data Loggers
Multichannel visualization.
  • 10.1" color touchscreen presents all 12 leads of data simultaneously
  • Widescreen display improves navigation, data entry and electrode verification
  • Supports pacemaker spike detection
  • Includes hook-up confirmation by detection and display of lead reversals
The ELI 280 Electrocardiograph provides comprehensive functionality in a portable, touchscreen device. The easy to use device is designed to meet the demands of high-volume ECG environments in both acute care and physician office environments. The widescreen data layout allows for easy menu navigation, fast entry of patient information and clear verification of proper electrode connections. It offers proven ECG performance, digital integration and VERITAS algorithm accuracy with more than 30 years of expertise for a more confident interpretation.