Metal Sieves
Supplier: United Scientific Supplies
UNMTSV02SL
470346-962PK 112.45 USD
470346-962
Metal Sieves
Sieves Test Sieves
Quickly sort particles of 4000 and 500 μm inside.
  • Stainless steel
  • Lid to contain dust
  • Aligned with GLOBE protocols
These stackable stainless steel mesh sieves are ideal for separating soil or sediment particles of multiple sizes. Available in six convenient sizes with a lid and catch pan. All sieves are 20 cm in diameter and 4 cm deep.