Bidirectional LED
Supplier: United Scientific Supplies
BILEDPK10
470346-960EA 0.8 USD
470346-960
Bidirectional LED
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Unique LED that works with any direction of current.
  • 2 pin connectors
  • Fits all standard bred boards
  • Colors are green and red
The Bidirectional LED has two colored bubs in one design. The direction of the current only illuminates one color at a time. This allows you to visually illustrate the direction of current in a direct current circuit. The bulb diameter is 5 mm.