Disposable Glass Pasteur Pipettes, 9"
Supplier: GLASSCO LABORATORY EQUIP SE
24673.043
470346-770CS 99 USD
470346-770
Disposable Glass Pasteur Pipettes, 9"
Pipettes Pasteur/Transfer Pipettes
Disposable pasteur pipettes are manufactured from borosilicate glass, and feature fire-polished tops and a straight-cut tip.
  • Ideal for use in a wide range of laboratory applications
  • Fire-polished tips facilitate bulb attachment
  • Borosilicate glass construction
Pasteur transfer pipettes are excellent for use in all general laboratory applications, including blood banks, hematology, chemistry, and bacteriology. Smooth, fire-polished tops facilitate bulb attachment. Glass pasteur pipettes have an inside diameter of 7 mm, which tapers to a straight-cut tip.More Product Information