Hausted APC Stretcher/Chair
Supplier: GUMBO MEDICAL
HAPCSC
470346-698EA 3489.09 USD
470346-698
Hausted APC Stretcher/Chair
Gurneys
Treat, transport and transfer.
  • Refurbished
  • 1 year parts warranty
  • For educational and training use only
Hausted’s All Purpose Chair (APC) is an easily adjustable patient transport and treatment device. The chair’s backrest can be positioned from sitting to supine and provides a stable working surface during patient treatment. Height positioning is hydraulically controlled using floor-level pedal, backrest and leg section adjustment is achieved with release levers at the rear of the patient backrest and at the foot of the unit. Four easy-rolling, steerable casters allow for maximum mobility and maneuverability. The APC is a multi-position chair and stretcher intended for intra- and inter- departmental transport of patients within a health care facility for patient treatment, transport or recovery.
Delivery information: This item has to ship by freight, additional shipping charges may apply.
Caution: This product is refurbished and is intended for educational and training use only.