Ohmeda Panda 3400 Infant Warmer
Supplier: GUMBO MEDICAL
OP3400
470346-672EA 3305.49 USD
470346-672
Ohmeda Panda 3400 Infant Warmer
Incubators Infant Incubators
Long term care warmer.
  • Refurbished
  • 1 year parts warranty
  • For educational and training use only
The Ohmeda Medical Panda™ Model 3400 Baby Warmer System provides a controlled source of radiant heat for infant and pediatric patients. The control system uses a microprocessor and provides both manual and servo modes of operation. The patient temperature, control temperature, and elapsed time displays are digital for ease of viewing. The control panel includes a one-hour elapsed timer with time displayed in minutes and seconds. The timer features optional audible tones for use during Apgar scoring. A complete audio and visual alarm system is included on the control panel. In addition, the Model 3400 has an elevating pedestal to raise and lower the bed-to-floor height. The Model 3400 is intended for surgical procedures and long-term care in the Nursery and L and D rooms.
Delivery information: This item has to ship by freight, Additional shipping charges may apply.
Caution: This product is refurbished and is intended for educational and training use only.