Medicine Dropper Pipettes
Supplier: GSC International
1300-12A
470346-558CS 70.4 USD
470346-558
Medicine Dropper Pipettes
Pipettes Pasteur/Transfer Pipettes
These eye dropper pipettes, also known as medicine droppers, are plastic and great for any lab.
  • 12 packs of 12
Crafted from durable plastic, these 3" droppers feature a 7 mm outside diameter and a 75 mm length, delivering a precise 1 ml capacity with each squeeze. Whether you need to transfer liquids or administer medication, these versatile pipettes get the job done efficiently. With 12 packs of 12 pipettes, you'll have a convenient supply of 144 droppers on hand for all your laboratory, medical, or everyday needs. Stop wasting time with inferior eye droppers - upgrade to our dependable medicine dropper pipettes and simplify your tasks today.