Quantum Levitation Starter Kit
Supplier: QUANTUM EXPERIENCE LTD
303-112
470346-496EA 716.39 USD
470346-496
Quantum Levitation Starter Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The starter kit is the fundamental, entry level kit for classroom demonstration of quantum levitation and flux pinning. Perfect for high-school students as well as university undergraduates.
  • The kit is easy to use
  • Highly durable and portable
A unique experimentation platform to demonstrate frictionless motion and quantum phenomena such as the Meissner effect and flux pinning.
Delivery information: The starter kit comes with handheld magnetic device, medium/large quantum levitator (various flavors available), plastic tweezers, and user manual.