Quantum Levitation Force Experiment
Supplier: QUANTUM EXPERIENCE LTD
410-112
470346-494EA 1172.29 USD
470346-494
Quantum Levitation Force Experiment
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Explore the magnetic forces between magnets, superconductors and metals.
  • The Meissner effect
  • Flux pinning
  • Magnetic forces
  • Letz' law
Visualize and measure the magnetic forces that Quantum Levitation so unique and unintuitive - see how a superconductor is repelled by a magnet (positive force - Meissner effect) and after it has been locked, it is attracted (negative forces - flux pinning) to it.